Novus Biologicals
Manufacturer Code:NBP274802JF646
Catalog # N274802J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G11 |
Immunogen | Full-length protein expressed in 293T cell transfected with human USP13 expression vector. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Deubiquitinating enzyme 13; Deubiquitinating enzyme 13 EC 3.1.2.15 EC 3.4.19.12 Isopeptidase T-3 ISOT-3 ISOT3IsoT-3 ubiquitin carboxyl-terminal hydrolase 13 ubiquitin specific peptidase 13 (isopeptidase T-3) ubiquitin specific protease 13 (isopeptidase T-3) ubiquitin thioesterase 13 Ubiquitin thiolesterase 13 Ubiquitin-specific-processing protease 13; Isopeptidase T-3; ISOT-3; Ubiquitin carboxyl-terminal hydrolase 13; ubiquitin specific protease 13 (isopeptidase T-3); Ubiquitin thioesterase 13; ubiquitin thiolesterase 13; Ubiquitin-specific-processing protease 13
Gene Aliases: IsoT-3; ISOT3; USP13
UniProt ID: (Human) Q92995
Entrez Gene ID: (Human) 8975
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.