Invitrogen
Manufacturer Code:PA572205
Catalog # PIPA572205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 387-420 amino acids from the Central region of DANRE usp13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 13; ISOT3 IsoT-3 deubiquitinating enzyme 13 isopeptidase T-3 ubiquitin carboxyl-terminal hydrolase 13 ubiquitin specific protease 13 (isopeptidase T-3) ubiquitin thioesterase 13 ubiquitin thiolesterase 13 ubiquitin-specific-processing protease 13; Ubiquitin carboxyl-terminal hydrolase 13; ubiquitin specific peptidase 13 (isopeptidase T-3); Ubiquitin thioesterase 13; Ubiquitin-specific-processing protease 13
Gene Aliases: usp13; wu:fc61g08
UniProt ID: (Zebrafish) F1QFS9
Entrez Gene ID: (Zebrafish) 558011
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.