Invitrogen
Manufacturer Code:PA523318
Catalog # PIPA523318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (522 REAEANRRPLPELVRAK 538) of USP13 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 13; Isopeptidase T-3; ISOT-3; ISOT3 IsoT-3 deubiquitinating enzyme 13 isopeptidase T-3 ubiquitin carboxyl-terminal hydrolase 13 ubiquitin specific protease 13 (isopeptidase T-3) ubiquitin thioesterase 13 ubiquitin thiolesterase 13 ubiquitin-specific-processing protease 13; Ubiquitin carboxyl-terminal hydrolase 13; ubiquitin specific peptidase 13 (isopeptidase T-3); ubiquitin specific protease 13 (isopeptidase T-3); Ubiquitin thioesterase 13; ubiquitin thiolesterase 13; Ubiquitin-specific-processing protease 13
Gene Aliases: 2700071E21Rik; AI848077; IsoT-3; ISOT3; USP13
UniProt ID: (Bovine) E1BMF7, (Human) Q92995, (Mouse) Q5BKP2
Entrez Gene ID: (Bovine) 100141289, (Horse) 100065844, (Human) 8975, (Dog) 478640, (Mouse) 72607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.