Invitrogen
Manufacturer Code:PA565023
Catalog # PIPA565023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human USP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 1; hUBP; ubiquitin carboxyl terminal hydrolase 1; Ubiquitin carboxyl-terminal hydrolase 1; Ubiquitin carboxyl-terminal hydrolase 1 Ubiquitin thioesterase 1 Ubiquitin-specific-processing protease 1 Deubiquitinating enzyme 1 hUBP; ubiquitin specific processing protease 1; ubiquitin specific protease 1; Ubiquitin thioesterase 1; ubiquitin thiolesterase 1; Ubiquitin-specific-processing protease 1
Gene Aliases: UBP; USP1
UniProt ID: (Human) O94782
Entrez Gene ID: (Human) 7398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.