Invitrogen
Manufacturer Code:PA544962
Catalog # PIPA544962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human USP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 1; hUBP; ubiquitin carboxyl terminal hydrolase 1; Ubiquitin carboxyl-terminal hydrolase 1; ubiquitin carboxyl-terminal hydrolase 1-like protein; Ubiquitin specific peptidase 1 UBP; ubiquitin specific processing protease 1; ubiquitin specific protease 1; Ubiquitin thioesterase 1; ubiquitin thiolesterase 1; Ubiquitin-specific-processing protease 1; usp1; usp1 {ECO:0000312|EMBL:AAH92574.1, ECO:0000312|RGD:1306461}
Gene Aliases: UBP; USP1
UniProt ID: (Bovine) Q29RP1, (Human) O94782, (Mouse) Q8BJQ2, (Rat) Q569C3
Entrez Gene ID: (Bovine) 516328, (Horse) 100070397, (Rabbit) 100358366, (Dog) 479549, (Human) 7398, (Pig) 100523071, (Guinea pig) 100722039, (Mouse) 230484, (Rat) 313387
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.