Invitrogen
Manufacturer Code:PA5110553
Catalog # PIPA5110553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 486-785 of human USP1 (NP_003359.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_3499 Deubiquitinating enzyme 1 hUBP Ubiquitin carboxyl-terminal hydrolase 1 Ubiquitin thioesterase 1 Ubiquitin-specific-processing protease 1 UBP Usp1; Deubiquitinating enzyme 1; hUBP; ubiquitin carboxyl terminal hydrolase 1; Ubiquitin carboxyl-terminal hydrolase 1; ubiquitin specific processing protease 1; ubiquitin specific protease 1; Ubiquitin thioesterase 1; ubiquitin thiolesterase 1; Ubiquitin-specific-processing protease 1; usp1
Gene Aliases: UBP; USP1
UniProt ID: (Human) O94782, (Mouse) Q8BJQ2
Entrez Gene ID: (Human) 7398, (Mouse) 230484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.