Proteintech
Manufacturer Code:219361AP150UL
Catalog # 50-173-3650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USH1G Fusion Protein Ag16571 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKS4A SANS USH1G Usher syndrome type 1G protein; Scaffold protein containing ankyrin repeats and SAM domain; Usher syndrome 1G (autosomal recessive); Usher syndrome type-1G protein
Gene Aliases: ANKS4A; SANS; USH1G
UniProt ID: (Human) Q495M9
Entrez Gene ID: (Human) 124590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.