Invitrogen
Manufacturer Code:PA131347
Catalog # PIPA131347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide DRKVAREFRHKVD corresponding to N terminal amino acids 2-14 of Human USH1C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIE-75 DFNB18 NY-CO-37 NY-CO-38 PDZ-45 PDZ-73 PDZ-73/NY-CO-38 PDZ73 ush1cpst; Antigen NY-CO-38/NY-CO-37; Autoimmune enteropathy-related antigen AIE-75; Harmonin; harmonin a1; PDZ domain-containing protein; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; Usher syndrome type-1C protein; Usher syndrome type-1C protein homolog
Gene Aliases: 2010016F01Rik; AIE-75; AIE75; DFNB18; DFNB18A; harmonin; NY-CO-37; NY-CO-38; PDZ-45; PDZ-73; PDZ-73/NY-CO-38; PDZ73; PDZD7C; USH1C; ush1cpst
UniProt ID: (Human) Q9Y6N9, (Bovine) Q3MHQ0, (Mouse) Q9ES64
Entrez Gene ID: (Human) 10083, (Bovine) 530709, (Rat) 308596, (Mouse) 72088
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.