Abnova
Manufacturer Code:H00007392M01A
Catalog # 89-105-711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5E9 |
Immunogen | USF2 (NP_0033581 a.a. &bsim 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant USF2.
For Research Use Only
Protein Aliases: bHLHb12; c-fos interacting protein; Class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; Upstream stimulatory factor 2; Upstream transcription factor 2
Gene Aliases: BHLHB12; FIP; USF2
UniProt ID: (Human) Q15853
Entrez Gene ID: (Human) 7392
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.