Invitrogen
Manufacturer Code:PA532637
Catalog # PIPA532637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to aa 327-346 of mouse USF-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb12; c-fos interacting protein; Class B basic helix-loop-helix protein 12; FIP; FIP bHLHb12 FOS-interacting protein c-fos interacting protein class B basic helix-loop-helix protein 12 major late transcription factor 2 upstream stimulatory factor 2; FOS-interacting protein; Major late transcription factor 2; Upstream stimulatory factor 2; Upstream transcription factor 2
Gene Aliases: BHLHB12; FIP; USF2
UniProt ID: (Human) Q15853
Entrez Gene ID: (Human) 7392
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.