Invitrogen
Manufacturer Code:PA5104472
Catalog # PIPA5104472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human USF2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb12; bHLHb12 c-fos interacting c-fos interacting protein class B basic helix-loop-helix protein 12 FIP FOS-interacting protein Major late transcription factor 2 transcription factor USF2 Upstream stimulatory factor 2 upstream transcription factor 2 upstream transcription factor 2 c-fos interacting Usf2 Usf-2; c-fos interacting protein; Class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; transcription factor USF2; Upstream stimulatory factor 2; Upstream transcription factor 2
Gene Aliases: BHLHB12; FIP; Usf-2; USF2
UniProt ID: (Human) Q15853, (Mouse) Q64705, (Rat) Q63665
Entrez Gene ID: (Human) 7392, (Mouse) 22282, (Rat) 81817
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.