Novus Biologicals
Manufacturer Code:NBP298839JF549
Catalog # NB055918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human USF1. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb11; BHLHB11 bHLHb11FCHL1 Class B basic helix-loop-helix protein 11 FCHL HYPLIP1 Major late transcription factor 1 MLTF MLTFI UEF upstream stimulatory factor 1 upstream transcription factor 1 USF; Class B basic helix-loop-helix protein 11; Major late transcription factor 1; Upstream stimulatory factor 1
Gene Aliases: BHLHB11; FCHL; FCHL1; HYPLIP1; MLTF; MLTFI; UEF; USF; USF1
UniProt ID: (Human) P22415
Entrez Gene ID: (Human) 7391
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.