Invitrogen
Manufacturer Code:PA541595
Catalog # PIPA541595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MDS032 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: embryonic development factor 2; p31; Protein D12; protein p31; putative MAPK activating protein PM26; Putative MAPK-activating protein PM26; Q-SNARE; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1 homolog; USE1-like protein; Vesicle transport protein USE1
Gene Aliases: 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; fj43b06; MDS032; P31; Q-snare; RGD1306660; SLT1; USE1; USE1L; wu:fj43b06; zgc:56402
UniProt ID: (Human) Q9NZ43, (Mouse) Q9CQ56, (Zebrafish) Q7ZTY7
Entrez Gene ID: (Horse) 100070008, (Human) 55850, (Pig) 100517081, (Bovine) 512890, (Dog) 609734, (Mouse) 67023, (Guinea pig) 100731909, (Rat) 290627, (Zebrafish) 393294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.