Invitrogen
Manufacturer Code:PA531265
Catalog # PIPA531265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 4 and 147 of Human USE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p31; P31 SLT1 Q-SNARE SNARE-like tail-anchored protein 1 homolog USE1-like protein protein p31 putative MAPK activating protein PM26 putative MAPK-activating protein PM26 vesicle transport protein USE1; protein p31; putative MAPK activating protein PM26; Putative MAPK-activating protein PM26; Q-SNARE; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1 homolog; USE1-like protein; Vesicle transport protein USE1
Gene Aliases: D12; MDS032; P31; SLT1; USE1; USE1L
UniProt ID: (Human) Q9NZ43
Entrez Gene ID: (Human) 55850
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.