Invitrogen
Manufacturer Code:PA5113496
Catalog # PIPA5113496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Vesicle transport protein USE1 protein (1-146AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010315L10Rik 5730403H22Rik AV002165 BOS_7155 D12 Ed2 embryonic development factor 2 fj43b06 hypothetical protein MGC56402 MDS032 p31 Protein D12 protein p31 Protein transport protein USE1 putative MAPK activating protein PM26 Putative MAPK-activating protein PM26 Q-snare RGD1306660 SLT1 SNAP receptor USE1 SNARE-like tail-anchored protein 1 SNARE-like tail-anchored protein 1 homolog Unconventional SNARE in the endoplasmic reticulum protein 1 unconventional SNARE in the ER 1 unconventional SNARE in the ER 1 homolog unconventional SNARE in the ER 1 homolog (S. cerevisiae) use1 Use1l USE1-like protein Vesicle transport protein USE1 wu:fj43b06 YGL098W zgc:56402; embryonic development factor 2; p31; Protein D12; protein p31; putative MAPK activating protein PM26; Putative MAPK-activating protein PM26; Q-SNARE; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1 homolog; USE1-like protein; Vesicle transport protein USE1
Gene Aliases: 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; MDS032; P31; Q-snare; SLT1; USE1; USE1L
UniProt ID: (Human) Q9NZ43, (Mouse) Q9CQ56
Entrez Gene ID: (Human) 55850, (Mouse) 67023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.