Novus Biologicals
Manufacturer Code:NBP303287100UL
Catalog # NB093957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 115-206 of human USAG1/SOSTDC1 (NP_056279.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cystine-knot containing secreted protein; cystine-knot containing secreted protein DKFZp564D206 Ectodermal BMP inhibitor ECTODIN sclerostin domain containing 1 sclerostin domain-containing protein 1 USAG-1 USAG1uterine sensitization-associated protein-1 Uterine sensitization-associated gene 1 protein; Ectodermal BMP inhibitor; Ectodin; Sclerostin domain-containing protein 1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein-1
Gene Aliases: CDA019; DAND7; ECTODIN; SOSTDC1; USAG1
UniProt ID: (Human) Q6X4U4
Entrez Gene ID: (Human) 25928
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.