R&D Systems
Manufacturer Code:AF7034SP
Catalog # AF7034SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human USAG1 Phe24-Ser206 Accession # Q6X4U4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CDA019 cystine-knot containing secreted protein DKFZp564D206 Ectodermal BMP inhibitor ECTODIN sclerostin domain containing 1 sclerostin domain-containing protein 1 SOSTDC1 USAG-1 USAG1uterine sensitization-associated protein-1 Uterine sensitization-associated gene 1 protein WISE; cystine-knot containing secreted protein; Ectodermal BMP inhibitor; Ectodin; Sclerostin domain-containing protein 1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein-1
Gene Aliases: CDA019; DAND7; ECTODIN; SOSTDC1; USAG1
UniProt ID: (Human) Q6X4U4
Entrez Gene ID: (Human) 25928
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.