Proteintech
Manufacturer Code:149131AP150UL
Catalog # 50-172-9829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UROS Fusion Protein Ag6711 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hydroxymethylbilane hydrolyase; Hydroxymethylbilane hydrolyase [cyclizing]; urogen III synthase; UROIIIS; UROIIIS uroporphyrinogen III synthase UROS; uroporphyrinogen III co-synthase; Uroporphyrinogen-III cosynthase; Uroporphyrinogen-III synthase
Gene Aliases: AI415298; UROIIIS; UROS; Uros3
UniProt ID: (Human) P10746, (Mouse) P51163
Entrez Gene ID: (Human) 7390, (Rat) 309070, (Mouse) 22276
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.