Invitrogen
Manufacturer Code:MA537497
Catalog # PIMA537497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 182CT27.2.2 |
Immunogen | URM1 recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2900073H19Rik C9orf74 MGC2668 RGD1306599 RP11-339B21.4 ubiquitin related modifier 1 ubiquitin related modifier 1 homolog ubiquitin related modifier 1 homolog (S. cerevisiae) ubiquitin-related modifier 1 ubiquitin-related modifier 1 homolog ubiquitin-related modifier URM1 URM1 YIL008W; ubiquitin related modifier 1 homolog; Ubiquitin-related modifier 1; ubiquitin-related modifier 1 homolog; URM1
Gene Aliases: C9orf74; URM1
UniProt ID: (Human) Q9BTM9
Entrez Gene ID: (Human) 81605
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.