Invitrogen
Manufacturer Code:PA567937
Catalog # PIPA567937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human URM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP11-339B21.4 C9orf74 MGC2668; ubiquitin related modifier 1 homolog; Ubiquitin-related modifier 1; ubiquitin-related modifier 1 homolog; URM1
Gene Aliases: 2900073H19Rik; C9orf74; RGD1306599; URM1
UniProt ID: (Human) Q9BTM9, (Mouse) Q9D2P4
Entrez Gene ID: (Human) 81605, (Mouse) 68205, (Rat) 311840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.