Invitrogen
Manufacturer Code:PA543630
Catalog # PIPA543630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human C19orf2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein NNX3; Protein phosphatase 1 regulatory subunit 19; protein phosphatase 1, regulatory subunit 19; RNA polymerase II subunit 5-mediating protein; RNA polymerase II, subunit 5-mediating protein; RPB5-mediating protein; Unconventional prefoldin RPB5 interactor; Unconventional prefoldin RPB5 interactor 1; Unconventional prefoldin RPB5 interactor-like protein; URI1 prefoldin-like chaperone; URI1 prefoldin-like chaperone FLJ10575 NNX3 RMP URI C19orf2 PPP1R19
Gene Aliases: C18H19orf2; C19orf2; C1H19orf2; C80913; NNX3; PPP1R19; RGD1310358; RMP; URI; URI1
UniProt ID: (Bovine) Q3B7M7, (Human) O94763, (Mouse) Q3TLD5
Entrez Gene ID: (Horse) 100053932, (Bovine) 515119, (Rabbit) 100352547, (Dog) 476501, (Human) 8725, (Mouse) 19777, (Guinea pig) 100717813, (Rat) 308537
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.