Proteintech
Manufacturer Code:149371AP150UL
Catalog # 50-172-9849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | URAT1 Fusion Protein Ag6770 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OAT4L OATL4 Renal specific transporter RST SLC22A12 URAT1 Urate anion exchanger 1; Organic anion transporter 4-like protein; Renal-specific transporter; RST; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; Solute carrier family 22 member 12; Urate anion exchanger 1; urate transporter 1
Gene Aliases: AI987855; OAT4L; OATL4; RST; SLC22A12; Slc22al2; UNQ6453/PRO34004; URAT1
UniProt ID: (Human) Q96S37, (Mouse) Q8CFZ5
Entrez Gene ID: (Pig) 100520540, (Human) 116085, (Mouse) 20521
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.