Invitrogen
Manufacturer Code:PA559761
Catalog # PIPA559761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UQCRFS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8 kDa subunit 9; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske mitochondrial Complex III subunit 5 Cytochrome b-c1 complex subunit 5 Rieske iron-sulfur protein RISP Ubiquinol-cytochrome c reductase iron-sulfur subunit Cytochrome b-c1 complex subunit 11 Complex III subunit IX Ubiquinol-cytochrome c reductase 8 kDa protein UQCRFS1; Cytochrome b-c1 complex subunit Rieske, mitochondrial; Liver regeneration-related protein LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; UQCRFS1 mitochondrial targeting sequence; UQCRFS1 MTS
Gene Aliases: LRRGT00195; RIP1; RIS1; RISP; UQCR5; UQCRFS1
UniProt ID: (Human) P47985, (Rat) P20788
Entrez Gene ID: (Human) 7386, (Rat) 291103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.