Invitrogen
Manufacturer Code:MA527789
Catalog # PIMA527789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1311 |
Immunogen | Recombit protein encompassing a sequence within the center region of human UQCRC1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Complex III subunit 1; Core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; ubiquinol-cytochrome c reductase core protein 1; Ubiquinol-cytochrome-c reductase complex core protein 1; UQCR1 D3S3191 ubiquinol-cytochrome c reductase core protein I QCR1
Gene Aliases: 1110032G10Rik; D3S3191; QCR1; UQCR1; UQCRC1
UniProt ID: (Human) P31930, (Rat) Q68FY0, (Mouse) Q9CZ13
Entrez Gene ID: (Human) 7384, (Rat) 301011, (Mouse) 22273
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.