Invitrogen
Manufacturer Code:PA582037
Catalog # PIPA582037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UQCRC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complex III subunit 1; Core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; D3S3191 QCR1 UQCR1; ubiquinol-cytochrome c reductase core protein 1; Ubiquinol-cytochrome-c reductase complex core protein 1
Gene Aliases: 1110032G10Rik; D3S3191; QCR1; UQCR1; UQCRC1
UniProt ID: (Human) P31930, (Rat) Q68FY0, (Mouse) Q9CZ13
Entrez Gene ID: (Human) 7384, (Rat) 301011, (Mouse) 22273
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.