Invitrogen
Manufacturer Code:PA588729
Catalog # PIPA588729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-111 of human UQCRB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complex III subunit 7; Complex III subunit VII; Cytochrome b-c1 complex subunit 7; mitochondrial ubiquinone-binding protein; QP-C; ubiquinol-cytochrome c reductase binding protein-like; Ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase, complex III subunit VI
Gene Aliases: 2210415M14Rik; MC3DN3; QCR7; QP-C; QPC; UQBC; UQBP; UQCR6; UQCRB; Uqcrbl; UQPC
UniProt ID: (Human) P14927
Entrez Gene ID: (Human) 7381, (Mouse) 67530, (Rat) 362897
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.