Novus Biologicals
Manufacturer Code:NBP162349
Catalog # NBP162349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to UCRC(ubiquinol-cytochrome c reductase complex (7.2 kD)) The peptide sequence was selected from the middle region of UCRC. Peptide sequence LFRRTSTFALTIIVGVMFFERAFDQGADAIYDHINEGKLWKHIKHKYENK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complex III subunit 9; Complex III subunit 9 Complex III subunit X cytochrome b-c1 complex subunit 9 Cytochrome c1 non-heme 7 kDa protein cytochrome C1 nonheme 7kDa protein HSPC051 HSPC151 QCR9 ubiquinol-cytochrome c reductase complex (7.2 kD) ubiquinol-cytochrome c reductase complex III subunit X ubiquinol-cytochrome c reductase complex III subunit X 7.2kDa UCCR7.2 UCRCUbiquinol-cytochrome c reductase complex 7.2 kDa protein; Complex III subunit X; Cytochrome b-c1 complex subunit 9; Cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; ubiquinol-cytochrome c reductase complex (7.2 kD); Ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa
Gene Aliases: HSPC051; HSPC119; HSPC151; QCR9; UCCR7.2; UCRC; UQCR10
UniProt ID: (Human) Q9UDW1
Entrez Gene ID: (Human) 29796
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.