Invitrogen
Manufacturer Code:MA516407
Catalog # PIMA516407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP73 |
Immunogen | Synthetic peptide corresponding to C-terminus of human uroplakin III protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: UP3a; UP3A UPIII UPIIIA UPK3 uroplakin III uroplakin-3a; UPIII; uroplakin 3; Uroplakin III; Uroplakin-3a
Gene Aliases: UP3A; UPIII; UPIIIA; UPK3; UPK3A
UniProt ID: (Human) O75631
Entrez Gene ID: (Human) 7380
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.