Abnova
Manufacturer Code:H00005976M01A
Catalog # 89-105-321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G3 |
Immunogen | UPF1 (NP_002902.21019 a.a. &bsim 1116 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant UPF1.
For Research Use Only
Protein Aliases: ATP-dependent helicase RENT1; delta helicase; FLJ43809; FLJ46894; HUPF1; KIAA0221; Nonsense mRNA reducing factor 1; NORF1; pNORF1; Regulator of nonsense transcripts 1; RENT1; smg-2 homolog, nonsense mediated mRNA decay factor; up-frameshift mutation 1 homolog; Up-frameshift suppressor 1 homolog; UPF1 regulator of nonsense transcripts homolog; yeast Upf1p homolog
Gene Aliases: HUPF1; KIAA0221; NORF1; pNORF1; RENT1; smg-2; UPF1
UniProt ID: (Human) Q92900
Entrez Gene ID: (Human) 5976
Molecular Function:
DNA binding protein
DNA helicase
RNA binding protein
RNA helicase
endoribonuclease
helicase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.