Invitrogen
Manufacturer Code:PA542183
Catalog # PIPA542183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human UPB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-alanine synthase; Beta-ureidopropionase; BUP-1; N-carbamoyl-beta-alanine amidohydrolase; Ureidopropionase beta BUP1; ureidopropionase, beta
Gene Aliases: AI195023; BUP1; UPB1; wu:fb69e03; zgc:64020
UniProt ID: (Human) Q9UBR1, (Rat) Q03248, (Mouse) Q8VC97
Entrez Gene ID: (Human) 51733, (Goat) 102183534, (Horse) 100053177, (Dog) 486398, (Bovine) 504557, (Rabbit) 100357276, (Rat) 116593, (Mouse) 103149, (Guinea pig) 100724141, (Zebrafish) 322660
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.