Invitrogen
Manufacturer Code:PA554855
Catalog # PIPA554855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UNK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RING finger protein unkempt homolog; unkempt homolog; ZC3H5 ZC3HDC5 RING finger protein unkempt homolog zinc finger CCCH domain-containing protein 5 zinc finger CCCH type domain containing 5 zinc finger CCCH-type containing 5 zinc finger CCCH-type domain containing 5; Zinc finger CCCH domain-containing protein 5; zinc finger CCCH type domain containing 5; zinc finger CCCH-type containing 5; zinc finger CCCH-type domain containing 5
Gene Aliases: KIAA1753; UNK; UNKEMPT; ZC3H5; ZC3HDC5
UniProt ID: (Human) Q9C0B0
Entrez Gene ID: (Human) 85451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.