Invitrogen
Manufacturer Code:PA569485
Catalog # PIPA569485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human UNCX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein unc-4 homolog; Homeobox protein unc-4 homolog Homeobox protein Uncx41 UNCX UNCX41; Homeobox protein Uncx4.1; Paired-type homeodomain transcription factor 1; truncated UNCX homeobox protein; UNC homeobox protein; Unc4.1 homeobox
Gene Aliases: Chx4; PHD1; UNCX; UNCX4.1
UniProt ID: (Human) A6NJT0, (Rat) P97830
Entrez Gene ID: (Human) 340260, (Rat) 29375, (Zebrafish) 798510
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.