Novus Biologicals
Manufacturer Code:NBP288558
Catalog # NB036180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Rat UNCX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein unc-4 homolog; homeobox protein unc-4 homolog homeobox protein Uncx4.1 UNC homeobox; Homeobox protein Uncx4.1; truncated UNCX homeobox protein; UNC homeobox protein; Unc4.1 homeobox
Gene Aliases: UNCX; UNCX4.1
UniProt ID: (Human) A6NJT0
Entrez Gene ID: (Human) 340260
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.