Invitrogen
Manufacturer Code:MA534800
Catalog # PIMA534800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG39-52 |
Immunogen | Recombinant protein within human SUN2 aa 500-700. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: B230369L08Rik BOS_6115 C030011B15 FRIGG KIAA0668 Protein unc-84 homolog B Rab5-interacting protein RAB5IP RGD1563141 Sad1/unc-84 protein-like 2 SUN domain-containing protein 2 Sun2 UNC84B; nuclear envelope protein; Protein unc-84 homolog B; Rab5-interacting protein; Rab5IP; Sad1 unc-84 domain protein 2; Sad1/unc-84 protein-like 2; SUN domain-containing protein 2; unc-84 homolog B
Gene Aliases: FRIGG; KIAA0668; RAB5IP; SUN2; UNC84B
UniProt ID: (Human) Q9UH99
Entrez Gene ID: (Human) 25777
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.