Invitrogen
Manufacturer Code:PA595741
Catalog # PIPA595741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human SUN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: nuclear envelope protein; Protein unc-84 homolog B; Rab5-interacting protein; Rab5IP; Sad1 unc-84 domain protein 2; Sad1/unc-84 protein-like 2; SUN domain-containing protein 2; unc-84 homolog B
Gene Aliases: B230369L08Rik; C030011B15; FRIGG; KIAA0668; RAB5IP; SUN2; UNC84B
UniProt ID: (Human) Q9UH99, (Mouse) Q8BJS4
Entrez Gene ID: (Human) 25777, (Mouse) 223697
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.