Novus Biologicals
Manufacturer Code:NBP29372020UL
Catalog # NB029304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SUN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FRIGG KIAA0668 MGC133055 MGC133056 nuclear envelope protein Protein unc-84 homolog B Rab5-interacting protein rab5IP Sad1 and UNC84 domain containing 2 Sad1 unc-84 domain protein 2 Sad1/unc-84 protein-like 2 SUN domain-containing protein 2 unc-84 homolog B unc-84 homolog B (C. elegans) UNC84B; nuclear envelope protein; Protein unc-84 homolog B; Rab5-interacting protein; Rab5IP; Sad1 unc-84 domain protein 2; Sad1/unc-84 protein-like 2; SUN domain-containing protein 2; unc-84 homolog B
Gene Aliases: FRIGG; KIAA0668; RAB5IP; SUN2; UNC84B
UniProt ID: (Human) Q9UH99
Entrez Gene ID: (Human) 25777
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.