R&D Systems
Manufacturer Code:MAB89661SP
Catalog # MAB89661SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1223A |
Immunogen | KLH-coupled N-terminal human UNC13D peptide Accession # Q70J99 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: FHL3 HLH3 HPLH3 MUNC13-4 Munc13-4protein unc-13 homolog D unc-13 homolog D (C. elegans); Munc13-4; Protein unc-13 homolog D
Gene Aliases: FHL3; HLH3; HPLH3; Munc13-4; UNC13D
UniProt ID: (Human) Q70J99
Entrez Gene ID: (Human) 201294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.