Invitrogen
Manufacturer Code:PA564015
Catalog # PIPA564015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UNC13B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of rat Munc13 (diacylglycerol-binding); munc13; MUNC13 UNC13 Unc13h2 homolog of rat Munc13 (diacylglycerol-binding) munc13-2 protein unc-13 homolog B; Munc13-2; Protein unc-13 homolog B
Gene Aliases: MUNC13; UNC13; UNC13B; Unc13h2
UniProt ID: (Human) O14795
Entrez Gene ID: (Human) 10497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.