Novus Biologicals
Manufacturer Code:NBP133037
Catalog # NBP133037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human UNC13B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hmunc13 homolog of rat Munc13 (diacylglycerol-binding) MGC133279 MGC133280 MUNC13 Munc13-2 protein unc-13 homolog B unc-13 homolog B (C. elegans) unc-13-like (C. elegans) UNC13Unc13h2; homolog of rat Munc13 (diacylglycerol-binding); munc13; Munc13-2; Protein unc-13 homolog B
Gene Aliases: MUNC13; UNC13; UNC13B; Unc13h2
UniProt ID: (Human) O14795
Entrez Gene ID: (Human) 10497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.