Proteintech
Manufacturer Code:550531AP150UL
Catalog # 50-173-5955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1032 Munc13 1 Protein unc 13 homolog A unc 13 homolog A (C. elegans) UNC13A; Munc13-1; Protein unc-13 homolog A; unc13a
Gene Aliases: 2410078G03Rik; KIAA1032; Munc13-1; UNC13A
UniProt ID: (Human) Q9UPW8, (Mouse) Q4KUS2
Entrez Gene ID: (Human) 23025, (Mouse) 382018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.