Invitrogen
Manufacturer Code:PA526380
Catalog # PIPA526380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 64-94 amino acids from the central region of human UNC119 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hRG4; HRG4 POC7 POC7A POC7 centriolar protein homolog A protein unc-119 homolog A retinal protein 4; mRG4; POC7 centriolar protein homolog A; Protein unc-119 homolog A; retinal gene 4; Retinal protein 4; unc-119 homolog
Gene Aliases: D11Bhm52; HRG4; IMD13; MRG4; POC7; POC7A; RG4; Rtg4; UNC119; Unc119h
UniProt ID: (Human) Q13432, (Mouse) Q9Z2R6
Entrez Gene ID: (Human) 9094, (Mouse) 22248
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.