Abnova
Manufacturer Code:PAB5396
Catalog # 89-131-843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human UNC119. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of UNC119.
For Research Use Only
Protein Aliases: HRG4; POC7 centriolar protein homolog A; Protein unc-119 homolog A; Retinal protein 4; unc-119 homolog
Gene Aliases: HRG4; IMD13; POC7; POC7A; RG4; UNC119
UniProt ID: (Human) Q13432
Entrez Gene ID: (Human) 9094
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.