Proteintech
Manufacturer Code:130651AP150UL
Catalog # 50-172-8461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UNC119 Fusion Protein Ag3757 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hRG4; HRG4 Protein unc 119 homolog A Retinal protein 4 RG4 unc 119 homolog (C. elegans) UNC119; mRG4; POC7 centriolar protein homolog A; Protein unc-119 homolog A; retinal gene 4; Retinal protein 4; rRG4; unc-119 homolog
Gene Aliases: CRG4; D11Bhm52; HRG4; IMD13; MRG4; POC7; POC7A; RG4; RRG4; Rtg4; UNC119; Unc119h; Uncl19
UniProt ID: (Human) Q13432, (Mouse) Q9Z2R6, (Rat) Q62885
Entrez Gene ID: (Dog) 491169, (Human) 9094, (Mouse) 22248, (Rat) 29402
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.