Invitrogen
Manufacturer Code:PA557559
Catalog # PIPA557559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human UMPS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ODC; OMPdecase; OPRT; OPRT OMPdecase OPRTase UMP synthase orotate phosphoribosyltransferase orotidine 5&prime-phosphate decarboxylase uridine 5&prime-monophosphate synthase; OPRTase; Orotate phosphoribosyltransferase; Orotidine 5'-phosphate decarboxylase; UMP synthase; Uridine 5'-monophosphate synthase; uridine monophosphate synthase
Gene Aliases: 1700095D23Rik; AA408257; AL033308; BB164745; OK/SW-cl.21; OPRT; UMPS
UniProt ID: (Human) P11172, (Mouse) P13439
Entrez Gene ID: (Human) 7372, (Mouse) 22247, (Rat) 288051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.