Proteintech
Manufacturer Code:113601AP150UL
Catalog # 50-172-7091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UMP/CMP kinase Fusion Protein Ag1920 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK; CK {ECO:0000255|HAMAP-Rule:MF_03172}; CMK CMPK CMPK1 Cytidine monophosphate kinase Cytidylate kinase Deoxycytidylate kinase UCK UMK UMP CMP kinase UMP CMPK UMP/CMP kinase UMP/CMPK UMPK Uridine monophosphate kinase; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidylate kinase; cytidylate kinase 1; dCMP kinase; dCMP kinase {ECO:0000255|HAMAP-Rule:MF_03172}; Deoxycytidylate kinase; deoxycytidylate kinase {ECO:0000255|HAMAP-Rule:MF_03172}; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); Nucleoside-diphosphate kinase; nucleoside-diphosphate kinase {ECO:0000255|HAMAP-Rule:MF_03172}; UMP-CMP kinase; UMP-CMP kinase 1; UMP-CMP kinase {ECO:0000255|HAMAP-Rule:MF_03172}; UMP/CMP kinase; UMP/CMP kinase {ECO:0000255|HAMAP-Rule:MF_03172}; UMP/CMPK; UMP/CMPK {ECO:0000255|HAMAP-Rule:MF_03172}; uridine monophosphate kinase; Uridine monophosphate/cytidine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase {ECO:0000255|HAMAP-Rule:MF_03172}
Gene Aliases: 0610011D08Rik; CK; CMK; CMPK; CMPK1; RGD1310116; UCK; UMK; UMP-CMPK; UMPK
UniProt ID: (Human) P30085, (Rat) Q4KM73, (Mouse) Q9DBP5
Entrez Gene ID: (Human) 51727, (Rat) 298410, (Mouse) 66588
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.