Invitrogen
Manufacturer Code:PA127859
Catalog # PIPA127859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fragment of yeast Transcriptional regulatory protein UME6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding transcriptional regulator UME6; Negative transcriptional regulator of IME2; Regulator of inducer of meiosis protein 16; Transcriptional regulatory protein UME6; Unscheduled meiotic gene expression protein 6
Gene Aliases: CAR80; CARGR1; NIM2; RIM16; UME6; YD8142.04C; YDR207C
UniProt ID: (Baker's yeast) P39001
Entrez Gene ID: (Baker's yeast) 851788
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.