Abnova
Manufacturer Code:H00009706M05
Catalog # 89-002-753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A12 |
Immunogen | ULK2 (AAH34988 743 a.a. ~ 843 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ULK2.
For Research Use Only
Protein Aliases: KIAA0623; Serine/threonine-protein kinase ULK2; Unc-51-like kinase 2; Unc51.2
Gene Aliases: ATG1B; KIAA0623; ULK2; Unc51.2
UniProt ID: (Human) Q8IYT8
Entrez Gene ID: (Human) 9706
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.