Invitrogen
Manufacturer Code:PA577808
Catalog # PIPA577808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the mid-protein of Human ULK2 (aa 558-569) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase ULK2 EC:2.7.11.1 Unc 51 (C. elegans) like kinase 2 ULK2&lowbarHUMAN KIAA0623 ATG1B Serine/threonine protein kinase ULK2 Unc-51-like kinase 2 Unc 51 like autophagy activating kinase 2 Unc51.2 ULK2 Unc 51 like kinase 2; Unc-51 like kinase 2; Unc-51-like kinase 2
Gene Aliases: ATG1B; KIAA0623; ULK2; Unc51.2
UniProt ID: (Human) Q8IYT8
Entrez Gene ID: (Human) 9706, (Rat) 303206
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.