Invitrogen
Manufacturer Code:PA5116106
Catalog # PIPA5116106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ULK2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A830085I22Rik ATG1B AU015340 EC:2.7.11.1 KIAA0623 mKIAA0623 Serine/threonine-protein kinase ULK2 serine/threonine-protein kinase Unc51.2 ULK2 unc-51 like autophagy activating kinase 2 unc-51 like kinase 2 Unc51.2 unc-51-like kinase 2; Serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; Unc-51 like kinase 2; Unc-51-like kinase 2
Gene Aliases: A830085I22Rik; ATG1B; AU015340; KIAA0623; mKIAA0623; ULK2; Unc51.2
UniProt ID: (Human) Q8IYT8, (Mouse) Q9QY01
Entrez Gene ID: (Human) 9706, (Mouse) 29869, (Rat) 303206
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.