Novus Biologicals
Manufacturer Code:NBP188881
Catalog # NBP188881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VVRRSNTSPMGFLRPGSCSPVPADTAQTVGRRLSTGSSRPYSPSPLVGTIPEQFSQCCCGHPQGHDSRSRNSSGSPVPQAQSPQSLLSGARLQSAPTLTDIYQNKQKLRKQHSDPVCPSHTGAGYSYSPQPSRPGSLGTSPTKH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATG1B EC 2.7.11 EC 2.7.11.1 KIAA0623serine/threonine-protein kinase ULK2 unc-51 (C. elegans)-like kinase 2 Unc51.2 Unc-51-like kinase 2 unc-51-like kinase 2 (C. elegans); Serine/threonine-protein kinase ULK2; Unc-51-like kinase 2
Gene Aliases: ATG1B; KIAA0623; ULK2; Unc51.2
UniProt ID: (Human) Q8IYT8
Entrez Gene ID: (Human) 9706
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.